Assay Details
| Gene Symbol: Gstk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644912.1 | XP_007643102.1 | 4 - 5 | 376 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gstk1| Gene Symbol: Gstk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644912.1 | XP_007643102.1 | 4 - 5 | 376 | 82 | |
Set Membership: |
|