Assay Details
| Gene Symbol: Mmp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647375.1 | XP_007645565.1 | 2 - 3 | 349 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp8| Gene Symbol: Mmp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647375.1 | XP_007645565.1 | 2 - 3 | 349 | 74 | |
Set Membership: |
|