Assay Details
| Gene Symbol: Sar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498491.2 | XP_003498539.1 | 2 - 3 | 185 | 98 | |
| XM_007628704.1 | XP_007626894.1 | 3 - 4 | 268 | 98 | ||
| GenBank mRNA | JI887013.1 | - | 2 - 3 | 173 | 98 | |
| JP043922.1 | - | 2 - 3 | 173 | 98 | ||
More Information
Set Membership: |
|

Sar1b
Genomic Map