Assay Details
| Gene Symbol: Fkbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499952.2 | XP_003500000.1 | 2 - 3 | 280 | 66 | |
| GenBank mRNA | JI872906.1 | - | 2 - 3 | 393 | 66 | |
| JP047257.1 | - | 2 - 3 | 393 | 66 | ||
More Information
Set Membership: |
|

Fkbp1a
Genomic Map