Assay Details
Gene Symbol: Commd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502148.2 | XP_003502196.1 | 1 - 2 | 219 | 85 | |
XM_007642786.1 | XP_007640976.1 | 1 - 2 | 220 | 85 | ||
GenBank mRNA | JI885260.1 | - | 1 - 2 | 194 | 85 | |
JP055993.1 | - | 1 - 2 | 194 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
