Assay Details
Gene Symbol: LOC100764363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503766.2 | XP_003503814.1 | 10 - 11 | 1577 | 99 | |
XM_007644324.1 | XP_007642514.1 | 9 - 10 | 1461 | 99 | ||
GenBank mRNA | JI868553.1 | - | 3 - 4 | 206 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
