Assay Details
| Gene Symbol: LOC100764363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503766.2 | XP_003503814.1 | 7 - 8 | 1168 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100764363| Gene Symbol: LOC100764363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503766.2 | XP_003503814.1 | 7 - 8 | 1168 | 98 | |
Set Membership: |
|