Assay Details
Gene Symbol: LOC100764363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503766.2 | XP_003503814.1 | 8 - 9 | 1404 | 98 | |
XM_007644324.1 | XP_007642514.1 | 7 - 8 | 1288 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
