Assay Details
Gene Symbol: LOC100764363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503766.2 | XP_003503814.1 | 9 - 10 | 1460 | 67 | |
XM_007644324.1 | XP_007642514.1 | 8 - 9 | 1344 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
