Assay Details
| Gene Symbol: Trib3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511497.2 | XP_003511545.1 | 3 - 4 | 793 | 67 | |
| XM_007652053.1 | XP_007650243.1 | 4 - 5 | 934 | 67 | ||
| XM_007652054.1 | XP_007650244.1 | 3 - 4 | 761 | 67 | ||
| XM_007652055.1 | XP_007650245.1 | 3 - 4 | 649 | 67 | ||
| GenBank mRNA | JI895402.1 | - | 2 - 3 | 587 | 67 | |
| JP063702.1 | - | 2 - 3 | 587 | 67 | ||
More Information
Set Membership: |
|

Trib3
Genomic Map