Assay Details
| Gene Symbol: Iah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514124.2 | XP_003514172.1 | 5 - 6 | 606 | 63 | |
| GenBank mRNA | JI872132.1 | - | 5 - 6 | 601 | 63 | |
More Information
Set Membership: |
|

Iah1
Genomic Map