Assay Details
| Gene Symbol: Plaur | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648992.1 | XP_007647182.1 | 307 | 97 | ||
| GenBank mRNA | AF172992.1 | - | 307 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plaur| Gene Symbol: Plaur | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648992.1 | XP_007647182.1 | 307 | 97 | ||
| GenBank mRNA | AF172992.1 | - | 307 | 97 | ||
Set Membership: |
|