Assay Details
| Gene Symbol: Mfhas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007620110.1 | XP_007618300.1 | 2 - 3 | 3038 | 70 | |
| GenBank mRNA | JI862825.1 | - | 2 - 3 | 636 | 70 | |
| JP045370.1 | - | 2 - 3 | 636 | 70 | ||
More Information
Set Membership: |
|

Mfhas1
Genomic Map