Assay Details
| Gene Symbol: Mapre2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641537.1 | XP_007639727.1 | 3 - 4 | 480 | 66 | |
| GenBank mRNA | JI869376.1 | - | 2 - 3 | 287 | 66 | |
More Information
Set Membership: |
|

Mapre2
Genomic Map