Assay Details
| Gene Symbol: LOC100761839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500581.2 | XP_003500629.1 | 4 - 5 | 418 | 56 | |
| XM_007641349.1 | XP_007639539.1 | 4 - 5 | 418 | 56 | ||
More Information
Set Membership: |
|

LOC100761839
Genomic Map