Assay Details
| Gene Symbol: Mgst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500908.2 | XP_003500956.1 | 2 - 3 | 323 | 67 | |
| XM_007641639.1 | XP_007639829.1 | 2 - 3 | 305 | 67 | ||
| XM_007641640.1 | XP_007639830.1 | 2 - 3 | 269 | 67 | ||
| GenBank mRNA | JI888647.1 | - | 1 - 2 | 151 | 67 | |
| JP038432.1 | - | 2 - 3 | 318 | 67 | ||
More Information
Set Membership: |
|

Mgst1
Genomic Map