Assay Details
| Gene Symbol: Klhl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502462.2 | XP_003502510.1 | 6 - 7 | 1635 | 62 | |
| XM_007643077.1 | XP_007641267.1 | 6 - 7 | 1732 | 62 | ||
| GenBank mRNA | JI864234.1 | - | 6 - 7 | 1618 | 62 | |
More Information
Set Membership: |
|

Klhl22
Genomic Map