Assay Details
| Gene Symbol: Paip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507308.2 | XP_003507356.1 | 2 - 3 | 310 | 101 | |
| XM_007647872.1 | XP_007646062.1 | 2 - 3 | 186 | 101 | ||
| GenBank mRNA | JP055798.1 | - | 2 - 3 | 309 | 101 | |
More Information
Set Membership: |
|

Paip2
Genomic Map