Assay Details
| Gene Symbol: Got2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509580.2 | XP_003509628.1 | 8 - 9 | 1128 | 59 | |
| GenBank mRNA | JI882271.1 | - | 8 - 9 | 1119 | 59 | |
| JP038917.1 | - | 8 - 9 | 1117 | 59 | ||
More Information
Set Membership: |
|

Got2
Genomic Map