Assay Details
| Gene Symbol: Pemt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643959.1 | XP_007642149.1 | 3 - 4 | 436 | 76 | |
| XM_007643960.1 | XP_007642150.1 | 3 - 4 | 436 | 76 | ||
| XM_007643961.1 | XP_007642151.1 | 2 - 3 | 328 | 76 | ||
More Information
Set Membership: |
|

Pemt
Genomic Map