Assay Details
| Gene Symbol: LOC100761790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652579.1 | XP_007650769.1 | 10 - 11 | 1348 | 75 | |
| GenBank mRNA | JI887762.1 | - | 7 - 8 | 701 | 75 | |
More Information
Set Membership: |
|

LOC100761790
Genomic Map