Assay Details
| Gene Symbol: LOC100761790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652579.1 | XP_007650769.1 | 7 - 8 | 1093 | 62 | |
| GenBank mRNA | JI887762.1 | - | 4 - 5 | 446 | 62 | |
More Information
Set Membership: |
|

LOC100761790
Genomic Map