Assay Details
| Gene Symbol: Bnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495047.2 | XP_003495095.2 | 6 - 7 | 811 | 74 | |
| XM_003495048.2 | XP_003495096.2 | 6 - 7 | 811 | 74 | ||
| GenBank mRNA | JI886284.1 | - | 6 - 7 | 729 | 74 | |
| JP056442.1 | - | 6 - 7 | 729 | 74 | ||
More Information
Set Membership: |
|

Bnip2
Genomic Map