Assay Details
| Gene Symbol: Ctsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495619.2 | XP_003495667.1 | 9 - 10 | 756 | 69 | |
| GenBank mRNA | JI895599.1 | - | 9 - 10 | 702 | 69 | |
More Information
Set Membership: |
|

Ctsh
Genomic Map