Assay Details
| Gene Symbol: Dnajb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501888.2 | XP_003501936.1 | 1 - 2 | 314 | 80 | |
| GenBank mRNA | JI872951.1 | - | 1 - 2 | 304 | 80 | |
| JP043469.1 | - | 1 - 2 | 304 | 80 | ||
More Information
Set Membership: |
|

Dnajb1
Genomic Map