Assay Details
| Gene Symbol: Ube2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502136.2 | XP_003502184.1 | 4 - 5 | 648 | 91 | |
| GenBank mRNA | JI889523.1 | - | 4 - 5 | 648 | 91 | |
| JP040318.1 | - | 4 - 5 | 648 | 91 | ||
More Information
Set Membership: |
|

Ube2e3
Genomic Map