Assay Details
| Gene Symbol: Pigg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502357.2 | XP_003502405.1 | 6 - 7 | 1289 | 72 | |
| XM_007642968.1 | XP_007641158.1 | 6 - 7 | 1289 | 72 | ||
| XM_007642969.1 | XP_007641159.1 | 5 - 6 | 844 | 72 | ||
| XM_007642970.1 | XP_007641160.1 | 6 - 7 | 1289 | 72 | ||
| XM_007642971.1 | XP_007641161.1 | 6 - 7 | 1289 | 72 | ||
| GenBank mRNA | JP038663.1 | - | 5 - 6 | 1230 | 72 | |
More Information
Set Membership: |
|

Pigg
Genomic Map