Assay Details
| Gene Symbol: LOC100766402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502567.2 | XP_003502615.1 | 12 - 13 | 2437 | 82 | |
| XM_007643191.1 | XP_007641381.1 | 12 - 13 | 2444 | 82 | ||
| XM_007643192.1 | XP_007641382.1 | 12 - 13 | 2580 | 82 | ||
| XM_007643193.1 | XP_007641383.1 | 13 - 14 | 2171 | 82 | ||
| GenBank mRNA | JI873561.1 | - | 2 - 3 | 221 | 82 | |
| JP048526.1 | - | 2 - 3 | 221 | 82 | ||
More Information
Set Membership: |
|

LOC100766402
Genomic Map