Assay Details
| Gene Symbol: LOC100766402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502567.2 | XP_003502615.1 | 1 - 2 | 479 | 74 | |
| XM_007643191.1 | XP_007641381.1 | 1 - 2 | 483 | 74 | ||
More Information
Set Membership: |
|

LOC100766402
Genomic Map