Assay Details
| Gene Symbol: LOC100766402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502567.2 | XP_003502615.1 | 2 - 3 | 571 | 117 | |
| XM_007643191.1 | XP_007641381.1 | 2 - 3 | 575 | 117 | ||
More Information
Set Membership: |
|

LOC100766402
Genomic Map