Assay Details
| Gene Symbol: LOC100766402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502567.2 | XP_003502615.1 | 4 - 5 | 1469 | 66 | |
| GenBank mRNA | JI893269.1 | - | 1 - 2 | 77 | 66 | |
More Information
Set Membership: |
|

LOC100766402
Genomic Map