Assay Details
| Gene Symbol: LOC100766402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502567.2 | XP_003502615.1 | 7 - 8 | 1729 | 68 | |
| XM_007643191.1 | XP_007641381.1 | 7 - 8 | 1736 | 68 | ||
| XM_007643192.1 | XP_007641382.1 | 7 - 8 | 1872 | 68 | ||
| XM_007643193.1 | XP_007641383.1 | 8 - 9 | 1463 | 68 | ||
| GenBank mRNA | JI893269.1 | - | 4 - 5 | 336 | 67 | |
More Information
Set Membership: |
|

LOC100766402
Genomic Map