Assay Details
| Gene Symbol: LOC100766402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502567.2 | XP_003502615.1 | 8 - 9 | 1788 | 75 | |
| XM_007643191.1 | XP_007641381.1 | 8 - 9 | 1795 | 75 | ||
| XM_007643192.1 | XP_007641382.1 | 8 - 9 | 1931 | 75 | ||
| XM_007643193.1 | XP_007641383.1 | 9 - 10 | 1522 | 75 | ||
| GenBank mRNA | JI893269.1 | - | 5 - 6 | 395 | 75 | |
More Information
Set Membership: |
|

LOC100766402
Genomic Map