Assay Details
| Gene Symbol: Batf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510718.2 | XP_003510766.1 | 2 - 3 | 280 | 64 | |
| XM_007651316.1 | XP_007649506.1 | 3 - 4 | 253 | 64 | ||
| GenBank mRNA | JI889171.1 | - | 2 - 3 | 208 | 64 | |
More Information
Set Membership: |
|

Batf2
Genomic Map