Assay Details
| Gene Symbol: LOC100770947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511880.2 | XP_003511928.2 | 13 - 14 | 1386 | 105 | |
| XM_007652378.1 | XP_007650568.1 | 14 - 15 | 1506 | 105 | ||
| XM_007652379.1 | XP_007650569.1 | 14 - 15 | 1428 | 105 | ||
More Information
Set Membership: |
|

LOC100770947
Genomic Map