Assay Details
| Gene Symbol: LOC100770947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511880.2 | XP_003511928.2 | 16 - 17 | 1626 | 76 | |
| XM_007652378.1 | XP_007650568.1 | 17 - 18 | 1746 | 76 | ||
| XM_007652379.1 | XP_007650569.1 | 17 - 18 | 1668 | 76 | ||
More Information
Set Membership: |
|

LOC100770947
Genomic Map