Assay Details
| Gene Symbol: LOC100770947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511880.2 | XP_003511928.2 | 19 - 20 | 1999 | 82 | |
| XM_007652378.1 | XP_007650568.1 | 20 - 21 | 2119 | 82 | ||
| XM_007652379.1 | XP_007650569.1 | 20 - 21 | 2041 | 82 | ||
More Information
Set Membership: |
|

LOC100770947
Genomic Map