Assay Details
| Gene Symbol: LOC100770947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511880.2 | XP_003511928.2 | 9 - 10 | 954 | 125 | |
| XM_007652378.1 | XP_007650568.1 | 9 - 10 | 954 | 125 | ||
| XM_007652379.1 | XP_007650569.1 | 9 - 10 | 954 | 125 | ||
| GenBank mRNA | JI872391.1 | - | 4 - 5 | 387 | 125 | |
| JP050227.1 | - | 4 - 5 | 387 | 125 | ||
More Information
Set Membership: |
|

LOC100770947
Genomic Map