Assay Details
| Gene Symbol: Fuom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512006.2 | XP_003512054.1 | 3 - 4 | 296 | 54 | |
| XM_007652516.1 | XP_007650706.1 | 3 - 4 | 428 | 54 | ||
More Information
Set Membership: |
|

Fuom
Genomic Map