Assay Details
| Gene Symbol: Hs6st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007625908.1 | XP_007624098.1 | 1 - 2 | 414 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hs6st1| Gene Symbol: Hs6st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007625908.1 | XP_007624098.1 | 1 - 2 | 414 | 80 | |
Set Membership: |
|