Assay Details
| Gene Symbol: Psmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644385.1 | XP_007642575.1 | 1 - 2 | 202 | 57 | |
| GenBank mRNA | JI894636.1 | - | 202 | 57 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmd8| Gene Symbol: Psmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644385.1 | XP_007642575.1 | 1 - 2 | 202 | 57 | |
| GenBank mRNA | JI894636.1 | - | 202 | 57 | ||
Set Membership: |
|