Assay Details
| Gene Symbol: LOC100754837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI865511.1 | - | 2 - 3 | 248 | 82 | |
| JP040752.1 | - | 2 - 3 | 248 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100754837| Gene Symbol: LOC100754837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI865511.1 | - | 2 - 3 | 248 | 82 | |
| JP040752.1 | - | 2 - 3 | 248 | 82 | ||