Assay Details
| Gene Symbol: LOC100754837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI885401.1 | - | 1 - 2 | 65 | 66 | |
| JP046187.1 | - | 1 - 2 | 65 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100754837| Gene Symbol: LOC100754837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI885401.1 | - | 1 - 2 | 65 | 66 | |
| JP046187.1 | - | 1 - 2 | 65 | 66 | ||
Set Membership: |
|