Assay Details
| Gene Symbol: Zbtb7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646989.1 | XP_007645179.1 | 5 - 6 | 1907 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb7a| Gene Symbol: Zbtb7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646989.1 | XP_007645179.1 | 5 - 6 | 1907 | 81 | |
Set Membership: |
|