Assay Details
| Gene Symbol: LOC100754837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652139.1 | XP_007650329.1 | 2 - 3 | 263 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100754837| Gene Symbol: LOC100754837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652139.1 | XP_007650329.1 | 2 - 3 | 263 | 63 | |
Set Membership: |
|