Assay Details
| Gene Symbol: Spata2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495076.2 | XP_003495124.1 | 3 - 4 | 352 | 67 | |
| XM_007651583.1 | XP_007649773.1 | 3 - 4 | 602 | 67 | ||
More Information
Set Membership: |
|

Spata2l
Genomic Map