Assay Details
| Gene Symbol: Nat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495673.2 | XP_003495721.1 | 3 - 4 | 372 | 59 | |
| XM_007643132.1 | XP_007641322.1 | 4 - 5 | 462 | 59 | ||
| XM_007643136.1 | XP_007641326.1 | 4 - 5 | 462 | 59 | ||
| XM_007643138.1 | XP_007641328.1 | 3 - 4 | 372 | 59 | ||
More Information
Set Membership: |
|

Nat9
Genomic Map