Assay Details
| Gene Symbol: Pole3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503457.2 | XP_003503505.1 | 2 - 3 | 308 | 66 | |
| GenBank mRNA | JI866871.1 | - | 2 - 3 | 284 | 66 | |
| JP050943.1 | - | 2 - 3 | 284 | 66 | ||
More Information
Set Membership: |
|

Pole3
Genomic Map