Assay Details
| Gene Symbol: LOC100757714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI864880.1 | - | 1 - 2 | 39 | 79 | |
| JP044415.1 | - | 1 - 2 | 39 | 79 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100757714| Gene Symbol: LOC100757714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI864880.1 | - | 1 - 2 | 39 | 79 | |
| JP044415.1 | - | 1 - 2 | 39 | 79 | ||