Assay Details
| Gene Symbol: LOC100757714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI864880.1 | - | 5 - 6 | 511 | 64 | |
| JP044415.1 | - | 5 - 6 | 511 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100757714| Gene Symbol: LOC100757714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI864880.1 | - | 5 - 6 | 511 | 64 | |
| JP044415.1 | - | 5 - 6 | 511 | 64 | ||
Set Membership: |
|